missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17F Antibody (8F5.1A9), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-12034CL1
This item is not returnable.
View return policy
Description
IL-17F Monoclonal antibody specifically detects IL-17F in Mouse samples. It is validated for Flow Cytometry
Specifications
| IL-17F | |
| Monoclonal | |
| CoraFluor 1 | |
| Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F | |
| This antibody was raised by immunising mice with murine IL-17F coupled to ovalbumin to generate a highly immunogenic complex. | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 112744 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| Flow Cytometry | |
| 8F5.1A9 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction