missing translation for 'onlineSavingsMsg'
Learn More

IL-17F Antibody (8F5.1A9), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18368922 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368922 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368922 Supplier Novus Biologicals Supplier No. NBP312034AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-17F Monoclonal antibody specifically detects IL-17F in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-17F
Applications Flow Cytometry
Classification Monoclonal
Clone 8F5.1A9
Conjugate Alexa Fluor 350
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F
Host Species Rabbit
Immunogen This antibody was raised by immunising mice with murine IL-17F coupled to ovalbumin to generate a highly immunogenic complex.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 112744
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.