missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17F Antibody (1058A), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB20571P
This item is not returnable.
View return policy
Description
IL-17F Monoclonal antibody specifically detects IL-17F in Mouse samples. It is validated for Flow Cytometry
Specifications
| IL-17F | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 1058A | |
| Flow Cytometry | |
| Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F | |
| E.coli-derived recombinant mouse IL-17F, Ala21-Val153, Accession # Q7TNI7 | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 112744 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction