missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17E/IL-25 Antibody (8B1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27324SS
This item is not returnable.
View return policy
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| IL-17E/IL-25 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 0.5-2ug/ml, Flow Cytometry 1-2ug/ml | |
| IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| 8B1 | |
| Unconjugated | |
| Q9H293 | |
| IL25 | |
| Full-length human IL17E recombinant protein was used as the immunogen for this antibody. | |
| 0.025 mg | |
| Cytokine Research | |
| 64806 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction