missing translation for 'onlineSavingsMsg'
Learn More

IL-17E/IL-25 Antibody (207710) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30737053 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737053 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737053 Supplier Novus Biologicals Supplier No. FAB1399JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-17E/IL-25 Monoclonal antibody specifically detects IL-17E/IL-25 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-17E/IL-25
Applications Western Blot
Classification Monoclonal
Clone 207710
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias IL17E, IL-17E, IL-25, interleukin 25, Interleukin-17E, interleukin-25
Host Species Rat
Immunogen E. coli-derived recombinant mouse IL-17E/IL-25, Val17-Ala169, Accession # NP_542767
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 64806
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.