missing translation for 'onlineSavingsMsg'
Learn More

IL-17B Mouse anti-Human, Clone: B-B58, Novus Biologicals™

Product Code. 18394723 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
Unit Size:
200µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394723 200 μg 200µL
18337662 500 μg 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18394723 Supplier Novus Biologicals Supplier No. NBP318115200UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-17B Monoclonal antibody specifically detects IL-17B in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-17B
Applications ELISA
Classification Monoclonal
Clone B-B58
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20
Host Species Mouse
Immunogen Recombinant human IL-17B. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells
Purification Method Affinity purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 27190
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.