missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-17A Recombinant Rabbit Monoclonal Antibody (217), APC
Rabbit Recombinant Monoclonal Antibody
£107.00 - £332.00
Specifications
| Antigen | IL-17A |
|---|---|
| Clone | 217 |
| Concentration | 0.1 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30534201
|
Invitrogen™
MA560516 |
100 Tests |
£332.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30536184
|
Invitrogen™
MA560517 |
25 Tests |
£107.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17A Recombinant Monoclonal Antibody for Flow
Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).Specifications
| IL-17A | |
| 0.1 mg/mL | |
| Flow Cytometry | |
| APC | |
| Rabbit | |
| RUO | |
| PBS with 0.5% BSA and 0.03% ProClin 300 | |
| Q62386 | |
| 16171 | |
| Recombinant Mouse IL-17A protein (Thr22-Ala158). | |
| Antibody |
| 217 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| IL17A | |
| Ctla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17a; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17A; interleukin-17; Interleukin17A; interleukin-17A; R-IL-17-A | |
| IL17A | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title