missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-17A Monoclonal Antibody (eBio64DEC17), Brilliant Violet™ 605, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£148.00 - £314.00
Specifications
| Antigen | IL-17A |
|---|---|
| Clone | eBio64DEC17 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18436578
|
Invitrogen™
406-7179-42 |
100 Tests |
£314.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18496488
|
Invitrogen™
406-7179-41 |
25 Tests |
£148.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17A Monoclonal antibody specifically detects IL-17A in Human samples. It is validated for Flow CytometrySpecifications
| IL-17A | |
| 5 μL/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q16552 | |
| 3605 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| eBio64DEC17 | |
| Flow Cytometry | |
| Brilliant Violet 605 | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Ctla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17a; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17A; interleukin-17; Interleukin17A; interleukin-17A; R-IL-17-A | |
| IgG1 κ | |
| Affinity chromatography | |
| IL17A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title