missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17/IL-17A Antibody (TC11-18H10), Alexa Fluor™ 700, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-72027AF700
This item is not returnable.
View return policy
Description
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IL-17/IL-17A | |
| Monoclonal | |
| 0.5 mg/ml | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml | |
| IL17A | |
| E. coli-expressed, recombinant mouse IL-17/IL-17A | |
| 0.1 mL | |
| Apoptosis, Cytokine Research, Immunology, Innate Immunity | |
| 3605 | |
| Mouse | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| TC11-18H10 | |
| Alexa Fluor 700 | |
| CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Recognizes mouse Interleukin 17, a 32kDa disulfide-linked homodimer consisting of two 136 amino acid chains, which belong to the IL-17 family (classified as IL-17A to IL-17F), and signal through the IL-17 receptor (IL-17R/CDw217). | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction