missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17/IL-17A Antibody (4k5F6), mFluor Violet 500 SE, Novus Biologicals™
Description
IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | IL-17/IL-17A |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 4k5F6 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A |
| Host Species | Mouse |
| Immunogen | A synthetic peptide between amino acids 1-75 of human IL-17A was used as the immunogen for the antibody. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?