missing translation for 'onlineSavingsMsg'
Learn More

IL-17/IL-17A Antibody (4k5F6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£158.00 - £418.00

Specifications

Antigen IL-17/IL-17A
Clone 4k5F6
Concentration 1.0 mg/mL
Dilution Western Blot 0.2 - 2 ug/mL
Applications Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18138991
View Documents
Novus Biologicals
NBP2-24765SS
0.025 mg
£158.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18787183
View Documents
Novus Biologicals
NBP2-24765
£418.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-17/IL-17A
1.0 mg/mL
Western Blot
Unconjugated
Mouse
Apoptosis, Cytokine Research, Immunology, Innate Immunity
Q6NZ94
3605
A synthetic peptide between amino acids 1-75 of human IL-17A was used as the immunogen for the antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
4k5F6
Western Blot 0.2 - 2 ug/mL
Monoclonal
Purified
RUO
Human
CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
IL17A
IgG2a κ
Protein G purified
The amino acid sequence used as immunogen is 100% homologous in Human, and 94% in Mouse, Rat, Monkey, Opossum, Horse, Cow and Chimpanzee S1PR1.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.