missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-16 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145465
This item is not returnable.
View return policy
Description
The protein encoded by IL-16 is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of IL-16 undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Alternate splicing results in multiple transcript variants.
Specifications
| IL-16 | |
| Polyclonal | |
| Unconjugated | |
| IL16 | |
| FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; Il16; IL-16; ILN; Interleukin; interleukin 16; Interleukin16; Interleukin-16; LCF; Lymphocyte chemoattractant factor; M-IL-16; mKIAA4048; neuronal interleukin 16; NIL16; PRIL16; prIL-16; prointerleukin 16; Pro-interleukin-16 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 3603, 374270 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q14005 | |
| IL16 | |
| Recombinant Chicken IL16 protein, Ser1183-Leu1454 (Accession #Q70XC4). | |
| 200 μL | |
| Primary | |
| Chicken, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction