missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-16 Monoclonal Antibody (55)
GREENER_CHOICE

Product Code. 30019471
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30019471 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30019471 Supplier Invitrogen™ Supplier No. MA552094

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-16 Monoclonal Antibody for IHC (P)

The protein encoded by IL-16 is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of IL-16 undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-16
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 55
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IL16
Gene Accession No. Q14005
Gene Alias FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; Il16; IL-16; ILN; Interleukin; interleukin 16; Interleukin16; Interleukin-16; LCF; Lymphocyte chemoattractant factor; M-IL-16; mKIAA4048; neuronal interleukin 16; NIL16; PRIL16; prIL-16; prointerleukin 16; Pro-interleukin-16
Gene Symbols IL16
Host Species Mouse
Immunogen Recombinant Human IL-16 Protein (Pro2-Ser130).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3603
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.