Learn More
Invitrogen™ IL-16 Monoclonal Antibody (14.1), Biotin
Description
M161B targets IL-16 in ELISA, and WB applications and shows reactivity with Human samples. The M161B immunogen is recombinant human IL-16. M161B detects IL-16 which has a predicted molecular weight of approximately 142 kDa. The M161B IL16 antibody (biotinylated conjugate of clone 14.1) has successfully been paired as the detection antibody in a sandwich ELISA with capture antibody M160E (clone 17.1) Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125 - 0.25 μg/mL for detection. Biotinylated antibody M161B (clone 14.1) and antibody M160E (clone 17.1) have successfully been used in combination with recombinant IL16 protein SIL16 in ELISA applications.
Specifications
Specifications
| Antigen | IL-16 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 14.1 |
| Concentration | 0.5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 4% BSA and Proprietary preservative |
| Gene | IL16 |
| Gene Accession No. | Q14005 |
| Gene Alias | FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; Il16; IL-16; ILN; Interleukin; interleukin 16; Interleukin16; Interleukin-16; LCF; Lymphocyte chemoattractant factor; M-IL-16; mKIAA4048; neuronal interleukin 16; NIL16; PRIL16; prIL-16; prointerleukin 16; Pro-interleukin-16 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.