missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-16 Antibody (70720), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3161MFV500
394.40 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IL-16 Monoclonal antibody specifically detects IL-16 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| IL-16 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Cytokine Research, Hypoxia, Immunology, Innate Immunity | |
| 3603 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 70720 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| FLJ16806, FLJ42735, FLJ44234, HsT19289, IL-16, interleukin 16 (lymphocyte chemoattractant factor), LCFprIL-16, lymphocyte chemoattractant factor, neuronal interleukin 16, NIL16, PRIL16, prointerleukin 16, pro-interleukin-16 | |
| E. coli-derived recombinant human IL-16 isoform 1, Met1203-Ser1332, Accession # Q14005 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction