missing translation for 'onlineSavingsMsg'
Learn More

IL-16 Antibody (70719) [Biotin], Novus Biologicals™

Product Code. 30736088 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736088 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736088 Supplier Novus Biologicals Supplier No. FAB316B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-16 Monoclonal antibody specifically detects IL-16 in Human samples. It is validated for Western Blot,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-16
Applications Western Blot, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 70719
Conjugate Biotin
Formulation PBS
Gene Alias FLJ16806, FLJ42735, FLJ44234, HsT19289, IL-16, interleukin 16 (lymphocyte chemoattractant factor), LCFprIL-16, lymphocyte chemoattractant factor, neuronal interleukin 16, NIL16, PRIL16, prointerleukin 16, pro-interleukin-16
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-16 isoform 1, Met1203-Ser1332, Accession # Q14005
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cytokine Research, Hypoxia, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3603
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.