missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-16 Antibody (14.1) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-09027-0.2mg
This item is not returnable.
View return policy
Description
IL-16 Monoclonal antibody specifically detects IL-16 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| IL-16 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 14.1 | |
| Unconjugated | |
| FLJ16806, FLJ42735, FLJ44234, HsT19289, IL-16, interleukin 16 (lymphocyte chemoattractant factor), LCFprIL-16, lymphocyte chemoattractant factor, neuronal interleukin 16, NIL16, PRIL16, prointerleukin 16, pro-interleukin-16 | |
| The antibody was raised against IL-16 using a synthetic peptide corresponding to the predicted amino acid sequence of a hydrophilic region of IL-16, conjugated to keyhole limpet hemocyanin. | |
| 0.2 mg | |
| Biologically Active Proteins, Cancer, Cytokine Research, Hypoxia, Immunology, Innate Immunity | |
| 3603 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction