missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-15R alpha Antibody (RM0103-6C38), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IL-15R alpha Monoclonal antibody specifically detects IL-15R alpha in Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Gene Alias | CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 |
| Host Species | Rat |
| Regulatory Status | RUO |
| Isotype | IgG2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?