Learn More
Invitrogen™ IL-15 Human Uncoated ELISA Kit with Plates
Specifically engineered for accurate and precise measurement of protein levels from samples including serum, plasma, and cell culture supernatants.
£174.00 - £392.00
Specifications
Target Species | Human |
---|---|
For Use With (Application) | ELISA |
Storage Requirements | 2°C to 8°C |
Synonym | Interleukin-15 |
Alert:
Contains Sodium azide
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15591167
|
Invitrogen™
88-7620-22 |
2 x 96 Tests |
£174.00
Pack of 2 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
15501177
|
Invitrogen™
88-7620-86 |
10 x 96 Tests |
£392.00
Pack of 10 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
Description
The Human Interleukin 15 (IL-15) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IL-15. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
IL-15 Human Uncoated ELISA Kit with Plates contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
Human | |
ELISA | |
2°C to 8°C | |
Interleukin-15 |
For Research Use Only