missing translation for 'onlineSavingsMsg'
Learn More

IL-15 Antibody (MM0368-13S34), DyLight 594, Novus Biologicals™

Product Code. 18421566 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421566 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421566 Supplier Novus Biologicals Supplier No. NBP211665DL594

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Western Blot, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-15
Applications Western Blot, Neutralization
Classification Monoclonal
Clone MM0368-13S34
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias IL-15MGC9721, interleukin 15, interleukin-15
Host Species Mouse
Immunogen Human recombinant IL-15
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3600
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.