missing translation for 'onlineSavingsMsg'
Learn More

IL-15 Antibody (1048222) [CoraFluor™ 1], Novus Biologicals™

Product Code. 30731308 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30731308 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731308 Supplier Novus Biologicals Supplier No. FAB2472CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-15
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1048222
Conjugate CoraFluor 1
Formulation PBS
Gene Alias IL-15MGC9721, interleukin 15, interleukin-15
Host Species Mouse
Immunogen E. coli-derived human IL-15 protein, Asn49-Ser162, Accession # P40933.1
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3600
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.