missing translation for 'onlineSavingsMsg'
Learn More

IL-13R alpha 2 Antibody (MM0367-11S23), PE, Novus Biologicals™

Product Code. 18433681 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
18433681 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433681 Supplier Novus Biologicals Supplier No. NBP211663PE

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-13R alpha 2 Monoclonal antibody specifically detects IL-13R alpha 2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Conjugate PE
Formulation PBS
Immunogen Human recombinant IL-13R alpha 2 (extracellular domain)
Regulatory Status RUO
Gene ID (Entrez) 3598
Target Species Human
Form Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.