missing translation for 'onlineSavingsMsg'
Learn More

IL-13R alpha 2 Antibody (2E10), Novus Biologicals™

Product Code. 18338588 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338588 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338588 Supplier Novus Biologicals Supplier No. H00003598M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-13R alpha 2 Monoclonal antibody specifically detects IL-13R alpha 2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-13R alpha 2
Applications Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA, KnockDown
Classification Monoclonal
Clone 2E10
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000631
Gene Alias cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13R, IL-13R subunit alpha-2, IL-13RA2, IL-13R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein
Host Species Mouse
Immunogen IL13RA2 (NP_000631, 27 a.a. ~ 126 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DTEIKVNPPQDFEIVDPGYLGYLYLQWQPPLSLDHFKECTVEYELKYRNIGSETWKTIITKNLHYKDGFDLNKGIEAKIHTLLPWQCTNGSEVQSSWAET
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3598
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.