missing translation for 'onlineSavingsMsg'
Learn More

IL-13R alpha 2 Antibody (15), DyLight 680, Novus Biologicals™

Product Code. 18378340 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378340 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378340 Supplier Novus Biologicals Supplier No. NBP290385FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-13R alpha 2 Monoclonal specifically detects IL-13R alpha 2 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-13R alpha 2
Applications ELISA
Classification Monoclonal
Clone 15
Conjugate DyLight 680
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13R, IL-13R subunit alpha-2, IL-13RA2, IL-13R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-13R alpha 2 (NP_032382.1; Met 1-Lys 334).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3598
Test Specificity Has cross-reactivity in ELISA with: Human IL13RA2/ CD213A2 No cross-reactivity in ELISA with: Mouse IL13RA1/ CD213a1 Human CSF2RB/ CD131 Human IL4R/ IL4Ra/ CD124 Less than 10% cross-reactivity in ELISA with Human IL13RA1/ CD213a1
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.