missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-13R alpha 1 Rabbit anti-Mouse, Clone: 122, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90409-100UL
This item is not returnable.
View return policy
Description
IL-13R alpha 1 Monoclonal specifically detects IL-13R alpha 1 in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.
Specifications
| IL-13R alpha 1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3597 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 122 | |
| ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 | |
| bB128O4.2.1 (interleukin 13 receptor, alpha 1), Cancer/testis antigen 19, CD213A1, CD213a1 antigen, CT19, IL13 receptor alpha-1 chain, IL-13 receptor subunit alpha-1, IL13R, IL-13R subunit alpha-1, IL-13Ra, IL13RA, IL-13RA1, IL-13R-alpha-1, interleukin 13 receptor, alpha 1, interleukin-13 receptor subunit alpha-1, NR4 | |
| Recombinant Mouse IL-13R alpha 1 protein | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction