missing translation for 'onlineSavingsMsg'
Learn More

IL-13R alpha 1 Rabbit anti-Human, Clone: 26, Novus Biologicals™

Rabbit Monoclonal Antibody

£286.00 - £429.00

Specifications

Antigen IL-13R alpha 1
Clone 26
Dilution ELISA 1:5000-1:10000
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18231428
View Documents
Novus Biologicals
NBP2-89716-50UL
50 μg
£286.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18276926
View Documents
Novus Biologicals
NBP2-89716-100UL
100 μg
£429.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-13R alpha 1 Monoclonal specifically detects IL-13R alpha 1 in Human samples. It is validated for ELISA.
Specifications

Specifications

IL-13R alpha 1
ELISA 1:5000-1:10000
Monoclonal
Purified
RUO
0.2 um filtered solution in PBS
3597
IgG
Protein A purified
26
ELISA
Unconjugated
Rabbit
Human
bB128O4.2.1 (interleukin 13 receptor, alpha 1), Cancer/testis antigen 19, CD213A1, CD213a1 antigen, CT19, IL13 receptor alpha-1 chain, IL-13 receptor subunit alpha-1, IL13R, IL-13R subunit alpha-1, IL-13Ra, IL13RA, IL-13RA1, IL-13R-alpha-1, interleukin 13 receptor, alpha 1, interleukin-13 receptor subunit alpha-1, NR4
Recombinant Human IL-13R alpha 1 protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.