missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL-13R alpha 1 Antibody (MK-6105) - IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28311-50ug
This item is not returnable.
View return policy
Description
IL-13R alpha 1 Monoclonal antibody specifically detects IL-13R alpha 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-13R alpha 1 | |
| Monoclonal | |
| Unconjugated | |
| P78552 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| MK-6105 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| bB128O4.2.1 (interleukin 13 receptor, alpha 1), Cancer/testis antigen 19, CD213A1, CD213a1 antigen, CT19, IL13 receptor alpha-1 chain, IL-13 receptor subunit alpha-1, IL13R, IL-13R subunit alpha-1, IL-13Ra, IL13RA, IL-13RA1, IL-13R-alpha-1, interleukin 13 receptor, alpha 1, interleukin-13 receptor subunit alpha-1, NR4 | |
| IL-13Ra1/ CD213a1 | |
| 50 μg | |
| CD Markers, Immunology, Innate Immunity, Signal Transduction | |
| 3597 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction