missing translation for 'onlineSavingsMsg'
Learn More

PeproTech IL-13 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  PeproTech 500-P224BT-50UG

 View more versions of this product

Product Code. 17835773

  • £228.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: TPGPVRRSTS PPVALRELIE ELSNITQDQK TSLCNSSMVW SVDLTAGGFC AALESLTNIS SCNAIHRTQR ILNGLCNQKA SDVASSPPDT KIEVAQFISK LLNYSKQLFR YGH. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat IL-13. Anti-Rat IL-13-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Rat IL-13 by sandwich ELISA (using 100 mL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti- Rat IL-13 (500-P224) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat IL-13. Western Blot: To detect Rat IL-13 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 mg/mL. When used in conjunction with compatible development reagents the detection limit for Recombinant Rat IL-13 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P224BT-1MG will be provided as 2 x 500 μg

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-13
Polyclonal
Biotin
Il13
ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600
Rabbit
Antigen affinity chromatography
RUO
116553
-20°C
Lyophilized
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
P42203
Il13
E.coli-derived Recombinant Rat IL-13 (113 a.a.).
50 μg
Primary
Rat
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.