Learn More
Invitrogen™ IL-13 Monoclonal Antibody (PVM13-1), Functional Grade, eBioscience™

Mouse Monoclonal Antibody
£89.00
Specifications
| Antigen | IL-13 |
|---|---|
| Clone | PVM13-1 |
| Concentration | 1 mg/mL |
| Content And Storage | 4°C |
| Applications | Functional Assay, Immunostaining, Neutralization |
Description
Description: The PVM13-1 antibody reacts with human interleukin-13 (IL-13). IL-13, also known as NC30, is a 12.5 kDa protein secreted by activated T helper cells, CD8^+ T cells, and NK cells. The biological activities of IL-13 include suppression of macrophage cytotoxic activity, upregulation of IL-1RA expression and suppression of inflammatory cytokine expression. Applications Reported: The PVM13-1 antibody has been reported for use as the capture antibody in an IL-13 ELISPOT assay or ELISA, as well as for neutralization of IL-13 bioactivity. Applications Tested: The functional grade PVM13-1 antibody has been tested for ELISA and ELISPOT capture and neutralization of IL-13 bioactivity. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.Specifications
| IL-13 | |
| 1 mg/mL | |
| Functional Assay, Immunostaining, Neutralization | |
| Functional Grade | |
| Mouse | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| P35225 | |
| 3596 | |
| Primary | |
| Affinity Chromatography |
| PVM13-1 | |
| 4°C | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human | |
| Il13 | |
| ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 | |
| Il13 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.