Learn More
IL-13 Monoclonal Antibody (eBio13A), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 16-7133-050
Description
The eBio13A antibody reacts with mouse IL-13. IL-13 is a cytokine produced mainly by Th2 cells, but also by antigen-primed CD8 T cells. IL-13 has a strong involvement in allergic inflammation and parasitic clearing and in cancer models has been shown to have either inhibitory or stimulatory activity depending on the tumor. In humans, IL-13 is found to play a role in isotype switching in B cells. IL-13 is implicating in down modulating macrophage activity, through the reduction of pro-inflammatory cytokines (IL-1, IL-6, IL-8, IL-10, IL-12) Applications Reported: The eBio13A antibody has been reported useful for ELISA and ELISPOT capture, as well as intracellular staining for flow cytometric analysis.
IL-13 Monoclonal antibody specifically detects IL-13 in Mouse samples. It is validated for Control, ELISA, Functional Assay
Specifications
| IL-13 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| P20109 | |
| Rat | |
| 50 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 κ |
| ELISA, Functional Assay | |
| eBio13A | |
| Functional Grade | |
| Il13 | |
| ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; Interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 | |
| Affinity chromatography | |
| RUO | |
| 16163 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.