Learn More
Invitrogen™ IL-13 Monoclonal Antibody (eBio1316H), Biotin, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen™ 13-7135-85
Description
Description: The eBio1316H antibody reacts with mouse IL-13. The eBio1316H antibody is a neutralizing antibody. Mouse interleukin-13 (IL-13), originally designated p600, is a 11.6 kDa protein secreted by activated T helper cells, CD8+ T cells, and NK cells. IL-13 was originally identified in 1989 through differential hybridization of cDNA libraries prepared from T helper 1 and T helper 2 cells. IL-13 contains four cysteine residues, which form two intramolecular disulfide bonds. IL-13 shares many activities with IL-4; this relates to a requirement for IL-4Ralpha chain for signal transduction. The biological activities of IL-13 include suppression of macrophage cytotoxic activity, upregulation of IL-1RA expression, and suppression of inflammatory cytokine expression. IL-13 induces CD23 expression on B cells and enhances CD72, and expression of class II MHC gene products. Mouse IL-13 does not display the activities on T cells and B cell stimulatory effects which IL-4 exhibits. Mouse IL-13 is active on human and mouse cells. Applications Reported: The eBio1316H antibody has been reported useful for neutralization, ELISA detection, and ELISPOT detection. Applications Tested: The biotinylated eBio1316H antibody has been tested as the detection antibody in a sandwich ELISA (and ELISPOT assay) for analysis of mouse interleukin-13 (IL-13) in combination with the purified eBio13A (14-7133) antibody for capture and recombit mouse IL-13 (14-8131) as the standard.
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
Specifications
| IL-13 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P20109 | |
| Il13 | |
| Affinity chromatography | |
| RUO | |
| 16163 | |
| 4°C, do not freeze | |
| Liquid |
| ELISA | |
| eBio1316H | |
| Biotin | |
| Il13 | |
| ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 | |
| Rat | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.