missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL-13 Monoclonal antibody specifically detects IL-13 in Mouse samples. It is validated for Flow Cytometry, ELISA
Specifications
Specifications
| Antigen | IL-13 |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 13A |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Flow Cytometry, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 |
| Host Species | Rat |
| Immunogen | The immunogen for this antibody was IL13. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?