missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12 R beta 2 Antibody (305719), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB19592U
This item is not returnable.
View return policy
Description
IL-12 R beta 2 Monoclonal antibody specifically detects IL-12 R beta 2 in Human, Mouse samples. It is validated for Direct ELISA
Specifications
| IL-12 R beta 2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Direct ELISA | |
| 305719 | |
| Direct ELISA | |
| IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL-12R subunit beta-2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 2, Cys28-Asn622, Accession # Q99665 | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 3595 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction