missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IL-12 p70 ELISA Development Kit (TMB), PeproTech®
missing translation for 'orderingAttributeHoverText'
Quantity:
10 x 96 Tests
missing translation for 'unitSize'
Pack of 10
Beschreibung
IL-12 p70, also known as interleukin-12, is a cytokine that plays a critical role in the immune system's response to infections and cancer. It is produced by immune cells, including dendritic cells, macrophages, and B cells, in response to certain pathogens and tumor cells. IL-12 p70 stimulates the production of interferon-gamma and other cytokines, which promote the activation and proliferation of T cells and natural killer cells. IL-12 p70 has been studied as a potential therapeutic target for various diseases and conditions, including infectious diseases, autoimmune disorders, and cancer. It has been shown to enhance immune responses against certain infections and tumors, and to modulate inflammatory responses in autoimmune diseases.
Spezifikation
Spezifikation
| Accession Number | P43431, P43432 |
| Assay Range | Lot-specific |
| Conjugate | HRP |
| Product Type | ELISA |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 16159, 16160 |
| Gene Symbol | IL12A, Il12b |
| Kit Contents | Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate |
| Quantity | 10 x 96 Tests |
| Regulatory Status | RUO |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur