missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12 p70/IL-12A Antibody (2Y37), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22038PE
This item is not returnable.
View return policy
Description
IL-12 p70/IL-12A Monoclonal specifically detects IL-12 p70/IL-12A in Human samples. It is validated for Flow Cytometry.
Specifications
| IL-12 p70/IL-12A | |
| Monoclonal | |
| PE | |
| P29459 | |
| IL12A | |
| Human recombinant IL-12 p70/IL-12A | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 3592 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 2Y37 | |
| Flow Cytometry | |
| CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction