missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12 p70/IL-12A Antibody (2Y37) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£171.00 - £437.00
Specifications
| Antigen | IL-12 p70/IL-12A |
|---|---|
| Clone | 2Y37 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:10-1:20, CyTOF-ready |
| Applications | Western Blot, CyTOF, Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18125484
|
Novus Biologicals
NBP2-22038-0.025MG |
0.025 mg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18130112
|
Novus Biologicals
NBP2-22038 |
0.1 mg | |||||||
Description
IL-12 p70/IL-12A Monoclonal specifically detects IL-12 p70/IL-12A in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| IL-12 p70/IL-12A | |
| LYOPH | |
| Western Blot, CyTOF, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Immunology, Innate Immunity | |
| P29459 | |
| 3592 | |
| IgG1 | |
| Protein G purified |
| 2Y37 | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:10-1:20, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
| IL12A | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title