Learn More
Invitrogen™ IL-12 p40 Monoclonal Antibody (K1E007_5H5), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | IL-12 p40 |
|---|---|
| Clone | K1E007_5H5 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30207495
|
Invitrogen™
MA555826 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30202089
|
Invitrogen™
MA556206 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: MCHQQLVISW FSLVFLASPL VAIWELKKDV YVVELDWYPD APGEMVVLTC DTPEEDGITW TLDQSSEVLG SGKTLTIQVK EFGDAGQYTC HKGGEVLSHS LLLLHKKEDG IWSTDILKDQ KEPKNKTFLR CEAKNYSGRF TCWWLTTIST DLTFSVKSSR GSSDPQGVTC GAATLSAERV RGDNKEYEYS VECQEDSACP AAEESLPIEV MVDAVHKLKY ENYTSSFFIR DIIKPDPPKN LQLKPLKNSR QVEVSWEYPD TWSTPHSYFS LTFCVQVQGK SKREKKDRVF TDKTSATVIC RKNASISVRA QDRYYSSSWS EWASVPCS
IL-12 p40 Monoclonal antibody specifically detects IL-12 p40 in Human samples. It is validated for Western BlotSpecifications
| IL-12 p40 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| CLMF; CLMF p35; CLMF p40; CLMF2; cytotoxic lymphocyte maturation factor 40 kDa subunit; Il12; IL12 p40; IL-12 p40 subunit; IL-12 subunit p35; IL-12 subunit p40; IL12, subunit p40; IL-12.p40; IL12A; IL-12A; Il12b; IL-12B; Il12p40; IL-12p40; IL-12p40 subunit; IMD28; IMD29; Interleukin; interleukin 12 p40 subunit; interleukin 12 subunit p40; interleukin 12, p40; interleukin 12B; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin-12 beta chain; interleukin-12 beta chain precursor; interleukin-12 p40; interleukin-12 p40 chain; interleukin-12 p40 subunit; interleukin-12 subunit beta; interleukin-12 subunit p40; interleukin-12p40; interleukin-23 p40 subunit; natural killer cell stimulatory factor, 40 kD subunit; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; p40; RP23-388G23.1 | |
| Recombinant human IL12B (immunogen range is 1-328). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Il12b |
| K1E007_5H5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P29460 | |
| 3593 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.