missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12/IL-35 p35 Antibody (B-T21), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14604AF350
This item is not returnable.
View return policy
Description
IL-12/IL-35 p35 Monoclonal antibody specifically detects IL-12/IL-35 p35 in Human samples. It is validated for ELISA
Specifications
| IL-12/IL-35 p35 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| CLMF;IL12A;IL-12A;IL-12p35;Ll12a;NFSK;NKSF1;P35 | |
| Recombinant human IL-12/IL-35 p35 | |
| 0.1 mL | |
| Innate Immunity | |
| Human | |
| Purified |
| ELISA | |
| B-T21 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction