missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12 / IL-23 p40 Rat, PE, Clone: C17.8, enQuireBio™
Rat Monoclonal Antibody
£99.15 - £246.00
Specifications
Antigen | IL-12 / IL-23 p40 |
---|---|
Clone | C17.8 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952648
|
EnquireBio
QAB96-PE-25UG |
25 μg |
£99.15
25µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942648
|
EnquireBio
QAB96-PE-100UG |
100 μg |
£246.00
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IL-12 / IL-23 p40 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
16160 | |
IgG2a κ | |
Protein A/G Purified |
C17.8 | |
Flow Cytometry | |
PE | |
RUO | |
NP_001290173 | |
Il12b | |
Primary | |
Store at 2-4°C. |