missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12/IL-23 p40 Antibody (B-P24), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14601JF646
This item is not returnable.
View return policy
Description
IL-12/IL-23 p40 Monoclonal antibody specifically detects IL-12/IL-23 p40 in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| IL-12/IL-23 p40 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40 | |
| Recombinant human IL-12/IL-23 p40 | |
| 0.1 mL | |
| Innate Immunity | |
| 3593 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA | |
| B-P24 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction