Learn More
PeproTech IL-11 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P01BT-25UG
Description
AA Sequence of recombinant protein: MPGPPPGPPR VSPDPRAELD STVLLTRSLL ADTRQLAAQL RDKFPADGDH NLDSLPTLAM SAGALGALQL PGVLTRLRAD LLSYLRHVQW LRRAGGSSLK TLEPELGTLQ ARLDRLLRRL QLLMSRLALP QPPPDPPAPP LAPPSSAWGG IRAAHAILGG LHLTLDWAVR GLLLLKTRL. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-11. Anti-Human IL-11-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human IL-11 by sandwich ELISA (using 100 μL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-11 (500-P01) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human IL-11 Western Blot: To detect Human IL-11 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible development reagents the detection limit for Recombinant Human IL-11 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P01BT-1MG will be provided as 2 x 500 μg
Interleukins (ILs) are a large group of cytokines that are produced mainly by leukocytes, although some are produced by certain phagocytes and auxiliary cells. ILs have a variety of functions, but most function to direct other immune cells to divide and differentiate. Each IL acts on a specific, limited group of cells through a receptor specific for that IL.
Specifications
| IL-11 | |
| Polyclonal | |
| Biotin | |
| IL11 | |
| adipogenesis inhibitory factor; AGIF; il 11; IL10RA; Il11; IL-11; IL11 inhibitor; IL-11ra; ILN; Interleukin; interleukin 11; Interleukin11; interleukin-11; MGC2146; Oprelvekin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3589 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P20809 | |
| IL11 | |
| E.coli-derived Recombinant Human IL-11. | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.