missing translation for 'onlineSavingsMsg'
Learn More

IL-10R alpha Rat anti-Mouse, Clone: 1B1.3a, Novus Biologicals™

Product Code. 18342023 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342023 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342023 Supplier Novus Biologicals Supplier No. NBP30733020UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-10R alpha Monoclonal specifically detects IL-10R alpha in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10R alpha
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 1B1.3a
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 0.5-1 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha
Host Species Rat
Immunogen Purified recombinant extracellular region of Mouse IL-10R alpha
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3587
Target Species Mouse
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.