missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10R alpha Antibody (OTI1D10), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71030AF488
This item is not returnable.
View return policy
Description
IL-10R alpha Monoclonal specifically detects IL-10R alpha in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| IL-10R alpha | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IL10RA | |
| Full length recombinant protein of human IL-10 R alpha (NP_001549) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1D10 | |
| Western Blot, Immunohistochemistry | |
| CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha | |
| Mouse | |
| Affinity Purified | |
| Cytokine Research | |
| 3587.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction