missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10R alpha Antibody (1B1.3a), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08221CL1
This item is not returnable.
View return policy
Description
IL-10R alpha Monoclonal antibody specifically detects IL-10R alpha in Human (Negative),Mouse samples. It is validated for Flow Cytometry,Immunocytochemistry/Immunofluorescence
Specifications
| IL-10R alpha | |
| Monoclonal | |
| CoraFluor 1 | |
| CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha | |
| Purified recombinant extracellular region of Mouse IL-10R alpha (Uniprot: Q61727) | |
| 0.1 mL | |
| Cytokine Research | |
| 3587 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| 1B1.3a | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human (Negative), Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction