missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10 Rat anti-Human, Virus, BB700, Clone: JES3-19F1, BD Horizon™
Rat Monoclonal Antibody
Brand: BD Biosciences 566567
This item is not returnable.
View return policy
Description
IL-10 Monoclonal antibody specifically detects IL-10 in Human,Virus samples. It is validated for Flow Cytometry.
Specifications
| IL-10 | |
| Monoclonal | |
| Brilliant Blue 700 (BB700) | |
| 100 Tests | |
| Primary | |
| Human, Virus | |
| IgG2a κ |
| Flow Cytometry | |
| JES3-19F1 | |
| Rat | |
| RUO | |
| IL-10 | |
| Refrigerate (2 to 8°C) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction