missing translation for 'onlineSavingsMsg'
Learn More

IL-10, clone: JES3-9D7, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-7108-81

Code : Z2

Additional Details : Weight : 0.00250kg

 View more versions of this product

Product Code. 15144067

  • £49.88 / 50µg


Estimated Shipment
30-07-2018
Add to basket

Description

Description

Description: The JES3-9D7 monoclonal antibody reacts with human interleukin-10 (IL-10).Applications Reported: The JES3-9D7 antibody has been reported for use in capture of human IL-10 by ELISA, intracellular flow cytometric analysis, and for neutralizing IL-10 bioactivity. (Use Functional Grade purified for functional studies cat. 16-7108).Applications Tested: The JES3-9D7 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of human Interleukin-10 (IL-10) in combination with the biotin JES3-12G8 (13-7109) antibody for detection and recombinant human IL-10 (14-8109) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1-4 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-10
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P22301
Interleukin-10, CSIF, IL-10, IL10A, TGIF
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
ELISAþDot blotþImmunohistochemistry
JES3-9D7
Unconjugated
IL10
Purified
IL10
IgG1, kappa
50μg
Store at 2-8°C.
Primary
3586
Videos

For Research Use Only.