missing translation for 'onlineSavingsMsg'
Learn More

IL-10 R beta Antibody (MM0360-8R24), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18422486 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
18422486 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422486 Supplier Novus Biologicals Supplier No. NBP211654MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-10 R beta Monoclonal antibody specifically detects IL-10 R beta in Human samples. It is validated for Western Blot, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10 R beta
Classification Monoclonal
Immunogen Human recombinant IL-10 R beta (extracellular domain)
Purification Method Protein G purified
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.