missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10 R beta Antibody (547324) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB53681JF635
This item is not returnable.
View return policy
Description
IL-10 R beta Monoclonal antibody specifically detects IL-10 R beta in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| IL-10 R beta | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL-10R2IL-10R subunit beta, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-10 Rβ, Ile21-Ser220, Accession # Q61190 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 547324 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity | |
| 3588 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction