missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL-10 R beta Monoclonal antibody specifically detects IL-10 R beta in Mouse samples. It is validated for ELISA, ELISA
Specifications
Specifications
| Antigen | IL-10 R beta |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 009 |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL-10R2IL-10R subunit beta, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-10 R beta (NP_032375.2; Met1-Ser222). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?