Learn More
IL-10 Monoclonal Antibody (JES5-2A5), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 16-7102-050
Description
The JES5-2A5 antibody reacts with mouse interleukin-10 (IL-10). The JES5-2A5 antibody is a neutralizing antibody. Mouse IL-10 is an 18 kDa factor also known as Cytokine Synthesis Inhibitory Factor (CSIF). In the mouse, Th2 cells, B1 cells, macrophages, and keratinocytes are the major cell subsets that produce IL-10. IL-10 inhibits synthesis of Th1 cytokines and proliferation of T cells, and acts as a costimulatory signal for mast cells, developing thymocytes and the Th2 response.
IL-10 Monoclonal antibody specifically detects IL-10 in Mouse samples. It is validated for Functional Assay, Neutralization
Specifications
| IL-10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| P18893 | |
| Rat | |
| 50 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 κ |
| Functional Assay, Neutralization | |
| JES5-2A5 | |
| Functional Grade | |
| IL10 | |
| CSIF; cytokine; cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; Interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF | |
| Affinity chromatography | |
| RUO | |
| 16153 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.